Bis(diphenylphosphino)methane, 97%, Thermo Scientific Chemicals
Bis(diphenylphosphino)methane, 97%, Thermo Scientific Chemicals
Bis(diphenylphosphino)methane, 97%, Thermo Scientific Chemicals
Bis(diphenylphosphino)methane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bis(diphenylphosphino)methane, 97%, Thermo Scientific Chemicals
CAS: 2071-20-7 | C25H22P2 | 384.40 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21112.0910 g
Catalog number ALFB21112.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
pH4.0-5.0
It is applied as an organic synthesis catalyst. It is also used as an intermediate in flavor and fragrances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied as an organic synthesis catalyst. It is also used as an intermediate in flavor and fragrances.
Solubility
Insoluble in water.
Notes
Air sensitive. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. R. J. Puddephatt. Chemistry of bis(diphenylphosphino)methane.Chem. Soc. Rev.,198312(2), 99-127.
  2. Gus J. Palenik, et al. Steric vs. electronic effects in palladium-thiocyanate complexes. Crystal structures of dithiocyanato[bis(diphenylphosphino)methane](II), isothiocyanatothiocyanato[1,2-bis(diphenylphosphino)ethane]palladium(II), and diisothiocyanato[1,3-bis(diphenylphosphino)propane]palladium(II).J. Am. Chem. Soc.,197597(5), 1059-1066.