Squalene, 98%, Thermo Scientific Chemicals
Squalene, 98%, Thermo Scientific Chemicals
Squalene, 98%, Thermo Scientific Chemicals
Squalene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Squalene, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFB20944.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSqualene
CAS111-02-4
Health Hazard 1H304
Health Hazard 3P301+P310-P331-P501c
Melting Point°C to 75°C
View more
Biosynthetic precursor to all steroids Squalene is used as a precursor to steroids and ergosterol and as an immunologic adjuvant in vaccines. It plays a vital role in the preparation of cholesterol, steroid hormones and vitamin D. It serves as a lubricant for fatty acid surfactant films adsorbed on iron oxide surfaces.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Biosynthetic precursor to all steroids Squalene is used as a precursor to steroids and ergosterol and as an immunologic adjuvant in vaccines. It plays a vital role in the preparation of cholesterol, steroid hormones and vitamin D. It serves as a lubricant for fatty acid surfactant films adsorbed on iron oxide surfaces.
Solubility
Miscible with diethyl ether and acetone. Immiscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Sánchez-Fidalgo, S.; Villegas, I.; Rosillo, M. Á.; Aparicio-Soto, M.; de la Lastra, C. A. Dietary squalene supplementation improves DSS-induced acute colitis by downregulating p38 MAPK and NFkB signaling pathways. Mol. Nutr. Food Res. 2015, 59 (2), 284-292.
  2. Kedl, J. D.; Kedl, R. M. How squalene GLAdly helps generate antigen-specific T cells via antigen-carrying neutrophils and IL-18. Eur. J. Immunol. 2015, 45 (2), 376-379.