Furazolidone, 98%, Thermo Scientific Chemicals
Furazolidone, 98%, Thermo Scientific Chemicals
Furazolidone, 98%, Thermo Scientific Chemicals
Furazolidone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Furazolidone, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Catalog number ALFB20834.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS450-91-9
IUPAC Name4-fluoro-2-methoxyaniline
Molecular FormulaC7H8FNO
InChI KeyBNRRMRUVYDETQC-UHFFFAOYSA-N
SMILESCOC1=C(N)C=CC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥94.0%
Monoamine oxidase inhibitor