2,4-Dichlorophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
2,4-Dichlorophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
2,4-Dichlorophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
2,4-Dichlorophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4-Dichlorophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
CAS: 6590-96-1 | C7H3Cl2NS | 204.07 g/mol
Have Questions?
Catalog NumberQuantity
ALFB20178.065 g
Catalog number ALFB20178.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS629-05-0
IUPAC Nameoct-1-yne
Molecular FormulaC8H14
InChI KeyUMIPWJGWASORKV-UHFFFAOYSA-N
SMILESCCCCCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4150 to 1.4170 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %