4-Sulfonamidophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
4-Sulfonamidophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
4-Sulfonamidophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
4-Sulfonamidophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Sulfonamidophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
CAS: 17852-52-7 | C6H10ClN3O2S | 223.675 g/mol
Have Questions?
Catalog NumberQuantity
ALFB20015.882.5 g
Catalog number ALFB20015.88
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 g
Request bulk or custom format
Chemical Identifiers
CAS3731-51-9
IUPAC Name1-(pyridin-2-yl)methanamine
Molecular FormulaC6H8N2
InChI KeyWOXFMYVTSLAQMO-UHFFFAOYSA-N
SMILESNCC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow to yellow
FormLiquid
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Refractive Index1.5435-1.5475 @ 20?C
It is a potentially useful intermediate in the production of Celebrex an anti inflammatory drug.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a potentially useful intermediate in the production of Celebrex an anti inflammatory drug.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from oxidizing agents, water/moisture and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
Frank Schönberger, et al.Simple and sensitive method for the determination of celecoxib in human serum by high-performance liquid chromatography with fluorescence detection.Journal of Chromatography B.2002,768(2), 255-260.