Sodium sulfate, anhydrous, 99%
Sodium sulfate, anhydrous, 99%
Sodium sulfate, anhydrous, 99%
Sodium sulfate, anhydrous, 99%
Thermo Scientific Chemicals
Sodium sulfate, anhydrous, 99%
CAS: 7757-82-6 | Na2O4S | 142.04 g/mol
Have Questions?
Catalog NumberQuantity
ALFA19890.36500 g
Catalog number ALFA19890.36
Price (MYR)
246.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
246.00
Add to cart
Specifications
Chemical Name or MaterialSodium sulfate
Melting Point884°C
Name Noteanhydrous
CAS7757-82-6
Recommended StorageAmbient temperatures
View more
Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.
Solubility
Soluble in water, glycerol and hydrogen iodide. Insoluble in ethanol and many organic solvents.
Notes
Anhydrous sodium sulfate is hygroscopic in nature, and absorbs moisture converting to the hydrated form. The hydrated form on heating gets converted back to anhydrous form.
RUO – Research Use Only