Cyclohexanone oxime, 97%, Thermo Scientific Chemicals
Cyclohexanone oxime, 97%, Thermo Scientific Chemicals
Cyclohexanone oxime, 97%, Thermo Scientific Chemicals
Cyclohexanone oxime, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexanone oxime, 97%, Thermo Scientific Chemicals
CAS: 100-64-1 | C6H11NO | 113.16 g/mol
Have Questions?
Catalog NumberQuantity
ALFA19820.36500 g
Catalog number ALFA19820.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS89-95-2
IUPAC Name(2-methylphenyl)methanol
Molecular FormulaC8H10O
InChI KeyXPNGNIFUDRPBFJ-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder or fused solid or clear liquid as melt
Melting Point (clear melt)33.0-39.0?C