Nerol, 97%, Thermo Scientific Chemicals
Nerol, 97%, Thermo Scientific Chemicals
Nerol, 97%, Thermo Scientific Chemicals
Nerol, 97%, Thermo Scientific Chemicals
Nerol, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALFA19808.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialNerol
CAS106-25-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c