p-Toluenesulfonic acid sodium salt, 90+%, Thermo Scientific Chemicals
p-Toluenesulfonic acid sodium salt, 90+%, Thermo Scientific Chemicals
p-Toluenesulfonic acid sodium salt, 90+%, Thermo Scientific Chemicals
p-Toluenesulfonic acid sodium salt, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenesulfonic acid sodium salt, 90+%, Thermo Scientific Chemicals
CAS: 657-84-1 | C7H7NaO3S | 194.18 g/mol
Have Questions?
Catalog NumberQuantity
ALFA19332.1850 g
Catalog number ALFA19332.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS12032-20-1
IUPAC Namedilutetium(3+) trioxidandiide
Molecular FormulaLu2O3
InChI KeyUGBIHFMRUDAMBY-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Lu+3].[Lu+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.