(3-Carboxypropyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals
(3-Carboxypropyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals
(3-Carboxypropyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals
(3-Carboxypropyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(3-Carboxypropyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals
CAS: 17857-14-6 | C22H22BrO2P | 429.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALFA19302.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS6968-76-9
IUPAC Name1-(3-methoxyphenyl)piperazine hydrochloridyl
Molecular FormulaC11H16ClN2O
InChI KeyUYZISVLMMYHXMS-UHFFFAOYSA-N
SMILES[Cl].COC1=CC=CC(=C1)N1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey to pink
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO4>=97.5 %
HPLC>=97.5 %
Reactant for preparation of Piperamide analogs as histone deacetylase (HDAC) inhibitors with antitumor activity, Boron-containing benzoxaboroles as antimalarial agents, Methyl alkenyl quinolones as antimycobacterial agents, Solandelactone E via Sharpless epoxidation, Taber cyclopropanation, chemoselective reductions and lithiation-borylation-allylation sequence, Organotin compounds as antifungal agents, Oleanolic acid derivatives as inhibitors of protein tyrosine phosphatase 1B for negative regulation of insulin pathway (a promising target for treatment of diabetes and obesity), 11-oxa prostaglandin analogs with ocular hypotensive activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reactant for preparation of Piperamide analogs as histone deacetylase (HDAC) inhibitors with antitumor activity, Boron-containing benzoxaboroles as antimalarial agents, Methyl alkenyl quinolones as antimycobacterial agents, Solandelactone E via Sharpless epoxidation, Taber cyclopropanation, chemoselective reductions and lithiation-borylation-allylation sequence, Organotin compounds as antifungal agents, Oleanolic acid derivatives as inhibitors of protein tyrosine phosphatase 1B for negative regulation of insulin pathway (a promising target for treatment of diabetes and obesity), 11-oxa prostaglandin analogs with ocular hypotensive activity.
Solubility
Soluble in water. soluble in ethanol.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
RUO – Research Use Only
General References:
  1. Yu Luo et. al. Synthesis and biological evaluation of piperamide analogues as HDAC inhibitors. Bioorganic & Medicinal Chemistry Letters,. 2011, 21 (16), 4844-4846 .
  2. Yong-Kang Zhang et. al. Synthesis and structure-activity relationships of novel benzoxaboroles as a new class of antimalarial agents. Bioorganic & Medicinal Chemistry Letters,. 2011, 21(2), 644-651 .