Sodium molybdenum oxide dihydrate, 98%, Thermo Scientific Chemicals
Sodium molybdenum oxide dihydrate, 98%, Thermo Scientific Chemicals
Sodium molybdenum oxide dihydrate, 98%, Thermo Scientific Chemicals
Sodium molybdenum oxide dihydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium molybdenum oxide dihydrate, 98%, Thermo Scientific Chemicals
CAS: 10102-40-6 | MoNa2O4 | 205.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA19222.36
Price (MYR)
1,238.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,238.00
Add to cart
Specifications
Chemical Name or MaterialSodium molybdenum oxide dihydrate
Melting Point100°C (in -2H2O)
CAS10102-40-6
Recommended StorageAmbient temperatures
RTECS NumberQA5085000
View more
Sodium molybdenum oxide is widely used as a fertilizer in agriculture, and a corrosion inhibitor being a non-oxidizing anodic inhibitor. It reduces galvanic corrosion potential, useful in industrial waste water treatment. It is recommended for treatment of whiptail in broccoli and cauliflower. It is a precursor for molybdenum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium molybdenum oxide is widely used as a fertilizer in agriculture, and a corrosion inhibitor being a non-oxidizing anodic inhibitor. It reduces galvanic corrosion potential, useful in industrial waste water treatment. It is recommended for treatment of whiptail in broccoli and cauliflower. It is a precursor for molybdenum.
Solubility
Soluble in water.
Notes
Stable. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Sun, K.; Feng, E.; Peng, H.; Ma, G.; Wu, Y.; Wang, H.; Lei, Z. A simple and high-performance supercapacitor based on nitrogen-doped porous carbon in redox-mediated sodium molybdate electrolyte. Electrochim. Acta 2015, 158, 361-367.
  2. Ebrahim, M. A.; Eidi, A.; Mortazavi, P.; Tavangar, S. M.; Tehrani, D. M. Hepatoprotective and antifibrotic effects of sodium molybdate in a rat model of bile duct ligation. J. Trace Elem. Med. Biol. 2015, 29, 242-248.