Methoxylamine hydrochloride, 98+%, Thermo Scientific Chemicals
Methoxylamine hydrochloride, 98+%, Thermo Scientific Chemicals
Methoxylamine hydrochloride, 98+%, Thermo Scientific Chemicals
Methoxylamine hydrochloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methoxylamine hydrochloride, 98+%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFA19188.065 g
Catalog number ALFA19188.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS14059-33-7
IUPAC Namebismuth(3+) vanadium tetraoxidandiide
Molecular FormulaBiO4V
InChI KeyHUUOUJVWIOKBMD-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[O--].[V].[Bi+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Methoxylamine hydrochloride is used as a reagent in the preparation of O-methyl oximes from aldehydes. It is orally bioavailable small molecule inhibitor with potential adjuvant activity. It is also used as internal standard for prostaglandin assays by gas chromatography-mass spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methoxylamine hydrochloride is used as a reagent in the preparation of O-methyl oximes from aldehydes. It is orally bioavailable small molecule inhibitor with potential adjuvant activity. It is also used as internal standard for prostaglandin assays by gas chromatography-mass spectrometry.
Solubility
Soluble in water, dimethyl sulfoxide and methanol.
Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and acid anhydrides.
RUO – Research Use Only
General References:
  1. Wachsmuth, C. J.; Hahn, T. A.; Oefner, P. J.; Dettmer, K. Enhanced metabolite profiling using a redesigned atmospheric pressure chemical ionization source for gas chromatography coupled to high-resolution time-of-flight mass spectrometry. Anal. Bioanal. Chem. 2015, 407 (22), 6669-6680.
  2. Lv, G.; Hu, D.; Cheong, K.; Li, Z.; Qing, X.; Zhao, J.; Li, S. Decoding glycome of Astragalus membranaceus based on pressurized liquid extraction, microwave-assisted hydrolysis and chromatographic analysis. J. Chromatogr. A 2015, 1409, 19-29.