Diisobutyl adipate, 97%, Thermo Scientific™
Diisobutyl adipate, 97%, Thermo Scientific™
Diisobutyl adipate, 97%, Thermo Scientific™
Diisobutyl adipate, 97%, Thermo Scientific™
Diisobutyl adipate, 97%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFA19186.1425 g
Catalog number ALFA19186.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDiisobutyl adipate
CAS141-04-8
Recommended StorageAmbient temperatures
Density0.954
RTECS NumberAV1480000
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.