1,4-Diaminoanthraquinone, tech. 90%, Thermo Scientific™
1,4-Diaminoanthraquinone, tech. 90%, Thermo Scientific™
1,4-Diaminoanthraquinone, tech. 90%, Thermo Scientific™
1,4-Diaminoanthraquinone, tech. 90%, Thermo Scientific™
1,4-Diaminoanthraquinone, tech. 90%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA19156.22
Price (MYR)
-
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material1,4-Diaminoanthraquinone
CAS128-95-0
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Use as the intermediate of synthetic dyes. Use anthraquinone reductive dyes and dispersal dyes and acid dyes and intermediates. Itself to disperse dyestuffs purple. 1,4-Diaminoanthraquinone oxidized with different metal atoms gives rise to conducting coordination polymers consisting of difunctional ligand molecules linked by metal atoms. 1, 4-diaminoanthraquinone has solar cell applications. A hybrid Si/poly (1, 4-diaminoanthraquinone) photoconductive diode for optical sensor applications. Acts as the sequential incorporation of the mediator in electrochemical sensing of glucose on a gold electrode.
Solubility
Soluble in benzene, pyridine, the difficulties, slightly soluble in hot aniline acetate, ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
General References:
  1. M. N. Vijayashree,; S. V. Subramanyam,; A. G. Samuelson. A new organic conducting material derived from 1,4-diaminoanthraquinone . Macromolecules,. 1992, 25 (11), 2988-2990.
  2. F. Brovelli,; B.L. Rivas,; J.C. Bernède,; M.A. del Valle,; F.R. Díaz,; Y. Berredjem. Electrochemical and optical studies of 1,4-diaminoanthraquinone for solar cell applications. Polymer Bulletin. 2007, 58 (3), 521-527.