Anthrone, 95%, Thermo Scientific Chemicals
Anthrone, 95%, Thermo Scientific Chemicals
Anthrone, 95%, Thermo Scientific Chemicals
Anthrone, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Anthrone, 95%, Thermo Scientific Chemicals
CAS: 90-44-8 | C14H10O | 194.23 g/mol
Have Questions?
Catalog NumberQuantity
ALFA19118.1425 g
Catalog number ALFA19118.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS174899-66-2
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear, colorless or yellow to red/brown
Assay (unspecified)>97.5%
Refractive Index1.4370-1.4410 @ 20?C
Water Content (Karl Fischer Titration)<0.5%
It finds its applications in organic synthesis. It is used for a popular cellulose assay and in the colorometric determination of carbohydrates in body fluids and of animal starch in liver tissue. The anthrones are used in pharmacy as laxative. They stimulate the motion of the colon and are responsible for less water reabsorption. They may only be used for a short amount of time, because long time use may lead to loss of electrolytes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in alcohol, benzene, and hot NaOH solutions. Insoluble in water
Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Asamenew G.; Bisrat D.; Mazumder A.; Asres K. In vitro antimicrobial and antioxidant activities of anthrone and chromone from the latex of Aloe harlana Reynolds.Phytotherapy research.2011,25(12), 1756-1760.
  2. Malterud KE.; Farbrot TL.; Huse AE.; Sund RB. Antioxidant and radical scavenging effects of anthraquinones and anthrones.Pharmacology.1993,47(1), 77-85.