2-Methyl-2-pentanol, 99%, Thermo Scientific Chemicals
2-Methyl-2-pentanol, 99%, Thermo Scientific Chemicals
2-Methyl-2-pentanol, 99%, Thermo Scientific Chemicals
2-Methyl-2-pentanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Methyl-2-pentanol, 99%, Thermo Scientific Chemicals
CAS: 590-36-3 | C6H14O | 102.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA19090.14
Price (MYR)
1,036.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,036.00
Add to cart
Specifications
Chemical Name or Material2-Methyl-2-pentanol
CAS590-36-3
Health Hazard 1H226-H315-H319-H335
Health Hazard 2GHS H Statement
H226-H319-H335
Flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
View more
It is used to study its antagonist activity. It reacts with a second target site to alter the spatial relation between the hydrophobic agonist binding site and the allosteric site. It may initiate the polymerization of isocyanates and epoxides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used to study its antagonist activity. It reacts with a second target site to alter the spatial relation between the hydrophobic agonist binding site and the allosteric site. It may initiate the polymerization of isocyanates and epoxides.
Solubility
Soluble in water at 20°C 32g/L. Soluble in alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. L. Perbellini.; F. Brugnone.; I. Pavan. New [3 + 4] Annulation. Identification of the metabolites of n-hexane, cyclohexane, and their isomers in men's urine.Toxicol. Appl. Pharmacol.1980,53(2), 220-229.
  2. Fumie Sato.; Hiroaki Ishikawa.; Masao Sato. Cp2TiCl2-catalyzed grignard exchange reactions with 1,3-dienes or styrenes. Preparation of allylic and α-arylethyl grignard reagents by a convenient and quantitative method.Tetrahedron Lett.1980,21(4), 365-368.