3'-Fluoro-4'-methoxyacetophenone, 99%, Thermo Scientific Chemicals
3'-Fluoro-4'-methoxyacetophenone, 99%, Thermo Scientific Chemicals
3'-Fluoro-4'-methoxyacetophenone, 99%, Thermo Scientific Chemicals
3'-Fluoro-4'-methoxyacetophenone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3'-Fluoro-4'-methoxyacetophenone, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number ALFA18972.14
Price (MYR)
860.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
860.00
Add to cart
Specifications
Chemical Name or Material3'-Fluoro-4'-methoxyacetophenone
CAS455-91-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c