2-Acrylamido-2-methylpropanesulfonic acid, 98%, Thermo Scientific Chemicals
2-Acrylamido-2-methylpropanesulfonic acid, 98%, Thermo Scientific Chemicals
2-Acrylamido-2-methylpropanesulfonic acid, 98%, Thermo Scientific Chemicals
2-Acrylamido-2-methylpropanesulfonic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Acrylamido-2-methylpropanesulfonic acid, 98%, Thermo Scientific Chemicals
CAS: 15214-89-8 | C7H13NO4S | 207.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1000 g
Catalog number ALFA18924.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Acrylamido-2-methylpropanesulfonic acid
CAS15214-89-8
Health Hazard 1H302-H312-H318
Health Hazard 2GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P363-P501c
View more
2-Acrylamide-2-methylpropanesulfonic acid has good complexion, adsorption, biological activity, surface activity, hydrolysis stability and thermal stability. It can be used in oil chemical, water treatment, synthetic fiber, printing and dyeing, plastics, water absorbing coatings, paper, bio-medical, magnetic materials and cosmetics industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Acrylamide-2-methylpropanesulfonic acid has good complexion, adsorption, biological activity, surface activity, hydrolysis stability and thermal stability. It can be used in oil chemical, water treatment, synthetic fiber, printing and dyeing, plastics, water absorbing coatings, paper, bio-medical, magnetic materials and cosmetics industries.
Solubility
Highly soluble in water (1×106 mg/L at 25°C ).
Notes
Hygroscopic. Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. George F. Fanta.; Robert C. Burr and William M. Doane. Graft polymerization of acryl amide and 2-acrylamido-2-methylpropanesulfonic acid onto starch.J. Appl. Polym. Sci.1979,24(9), 2015-2023.
  2. A. Pourjavadi.; Sh. Barzegar.; F. Zeidabadi. Synthesis and properties of biodegradable hydrogels of κ-carrageenan grafted acrylic acid-co-2-acrylamido-2-methylpropanesulfonic acid as candidates for drug delivery systems.React. Funct. Polym.2007,67(7), 644-654.