Sodium nitrite, 98%
Sodium nitrite, 98%
Sodium nitrite, 98%
Thermo Scientific Chemicals
Sodium nitrite, 98%
CAS: 7632-00-0 | NNaO2 | 69.00 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18668.36500 g
Catalog number ALFA18668.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS9002-93-1
IUPAC Name2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
InChI KeyJYCQQPHGFMYQCF-UHFFFAOYSA-N
SMILESCC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
Synonymtriton X polyoxyethylene (11) octyl phenyl ether polyoxyethylene (9) octylphenyl ether hydrol SW poletoxol peg-12 octyl phenyl ether triton X-102 ortho-gynol octoxynol octylphenoxypolyethoxyethanol
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to hazy
Cloud point53.0 to 67.0? (1% inwater)
Water Content (Karl Fischer Titration)≤ 0.50%
FormViscous liquid
Chromatographic Purity53.0 to 67.0? (1% inwater)
Sodium nitrite is used in many industrial applications viz. in meat curing, coloring and preserving. It is used as a reagent in analytical chemistry, an antidote in cyanide poisoning, an electrolyte in electrochemical grinding, as a cooling solution in closed loop systems and as an additive in industrial greases. It finds application as a corrosion inhibitor as well as in the rubber industry. In metallurgy, it is used for phosphatizing and detinning. It acts as a precursor to diazo dyes, nitroso compounds and various organic compounds like pharmaceuticals. As a food additive, it is used to prevent botulism.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium nitrite is used in many industrial applications viz. in meat curing, coloring and preserving. It is used as a reagent in analytical chemistry, an antidote in cyanide poisoning, an electrolyte in electrochemical grinding, as a cooling solution in closed loop systems and as an additive in industrial greases. It finds application as a corrosion inhibitor as well as in the rubber industry. In metallurgy, it is used for phosphatizing and detinning. It acts as a precursor to diazo dyes, nitroso compounds and various organic compounds like pharmaceuticals. As a food additive, it is used to prevent botulism.
Solubility
Soluble in water, methanol, ethanol, ammonia, nitric acid. Slightly soluble diethyl ether.
Notes
Decomposes above 320°. Hygroscopic. Incompatible with reducing agents, strong oxidizing agents, organics, finely powdered metals.
RUO – Research Use Only