Manganese(II) nitrate tetrahydrate, 98%, Thermo Scientific Chemicals
Manganese(II) nitrate tetrahydrate, 98%, Thermo Scientific Chemicals
Manganese(II) nitrate tetrahydrate, 98%, Thermo Scientific Chemicals
Manganese(II) nitrate tetrahydrate, 98%, Thermo Scientific Chemicals
Manganese(II) nitrate tetrahydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Manganese(II) nitrate tetrahydrate, 98%, Thermo Scientific Chemicals
CAS: 20694-39-7 | H8MnN2O10 | 251.006 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFA18521.30
Price (MYR)
424.00
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
424.00
Add to cart
Specifications
Chemical Name or MaterialManganese(II) nitrate tetrahydrate
Melting Point∼26°C
CAS20694-39-7
Health Hazard 1H272-H302-H314-H373
Health Hazard 2GHS H Statement
H272-H314-H318-H373-H302
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
View more
Manganese(II) nitrate tetrahydrate used as porcelain colorants. It is also used as a precursor for manganese dioxide and other manganese compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Manganese(II) nitrate tetrahydrate used as porcelain colorants. It is also used as a precursor for manganese dioxide and other manganese compounds.
Solubility
Soluble in ethanol and water.
Notes
Hygroscopic. Store in a cool place. Incompatible with strong reducing agents and strong acids.
RUO – Research Use Only
General References:
  1. Nkosi, F. P.; Jafta, C. J.; Kebede, M.; le Roux, L.; Mathe, M. K.; Ozoemena, K. I. Microwave-assisted optimization of the manganese redox states for enhanced capacity and capacity retention of LiAlxMn2- xO4 (x= 0 and 0.3) spinel materials. RSC Adv. 2015, 5 (41), 32256-32262.
  2. Baykal, A.; Güner, S.; Demir, A. Synthesis and magneto-optical properties of triethylene glycol stabilized Mn1-xZnxFe2O4 nanoparticles. J. Alloys Compd. 2015, 619, 5-11.