2-Naphthyl acetate, 99%, Thermo Scientific Chemicals
2-Naphthyl acetate, 99%, Thermo Scientific Chemicals
2-Naphthyl acetate, 99%, Thermo Scientific Chemicals
2-Naphthyl acetate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Naphthyl acetate, 99%, Thermo Scientific Chemicals
CAS: 1523-11-1 | C12H10O2 | 186.21 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18487.0910 g
Catalog number ALFA18487.09
Price (MYR)
339.00
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
339.00
Add to cart
Specifications
Chemical Name or Material2-Naphthyl acetate
CAS1523-11-1
Melting Point68°C to 70°C
Recommended StorageKeep cold
EINECS Number216-194-7