Nickel ammonium sulfate hexahydrate, 98%, Thermo Scientific Chemicals
Nickel ammonium sulfate hexahydrate, 98%, Thermo Scientific Chemicals
Nickel ammonium sulfate hexahydrate, 98%, Thermo Scientific Chemicals
Nickel ammonium sulfate hexahydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nickel ammonium sulfate hexahydrate, 98%, Thermo Scientific Chemicals
CAS: 7785-20-8 | H8N2NiO8S2·6H2O
Have Questions?
Catalog NumberQuantity
ALFA18441.36500 g
Catalog number ALFA18441.36
Price (MYR)
1,058.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,058.00
Add to cart
Specifications
Chemical Name or MaterialNickel ammonium sulfate hexahydrate
CAS7785-20-8
Health Hazard 1H302+H332-H317-H334-H341-H350i-H360D-H372
Health Hazard 2GHS H Statement
H334-H350-H360-H372-H341-H302-H332-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c
View more
Nickel ammonium sulfate hexahydrate is used as a nickel electrolyte for electroplating and as an analytical reagent. It is also used as a soil fertilizer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Nickel ammonium sulfate hexahydrate is used as a nickel electrolyte for electroplating and as an analytical reagent. It is also used as a soil fertilizer.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Meena, K.; Muthu, K.; Rajasekar, M.; Bhagavannarayana, G.; Meenakshisundaram, S. P.; Mojumdar, S. C. Urea/thiourea induced crystal growth of ammonium nickel sulfate hexahydrate and characterization studies. J. Therm. Anal. Calorim. 2015, 119 (2), 963-968.
  2. Singh, S.; Patel, A.; Prakashan, P. One pot oxidative esterification of aldehyde over recyclable cesium salt of nickel substituted phosphotungstate. Appl. Catal., A 2015, 505, 131-140.