Cobalt(II) acetate tetrahydrate, 98%
Cobalt(II) acetate tetrahydrate, 98%
Cobalt(II) acetate tetrahydrate, 98%
Cobalt(II) acetate tetrahydrate, 98%
Thermo Scientific Chemicals
Cobalt(II) acetate tetrahydrate, 98%
CAS: 6147-53-1 | C4H14CoO8 | 249.08 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18374.30250 g
Catalog number ALFA18374.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS51523-79-6
IUPAC Name(2,4-difluorophenyl)hydrazine
Molecular FormulaC6H6F2N2
InChI KeyRLZUIPTYDYCNQI-UHFFFAOYSA-N
SMILESNNC1=CC=C(F)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Cobalt(II) acetate tetrahydrate is used as an industrial catalyst to harden paints and varnishes, an active catalyst for oxidation and esterification reactions. It is also used to prepare metal complexes having unusual coordination geometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cobalt(II) acetate tetrahydrate is used as an industrial catalyst to harden paints and varnishes, an active catalyst for oxidation and esterification reactions. It is also used to prepare metal complexes having unusual coordination geometry.
Solubility
Soluble in water, alcohol, dilute acids and phenyl acetate.
Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and bases.
RUO – Research Use Only