Sudan III
Sudan III
Sudan III
Thermo Scientific Chemicals
Sudan III
CAS: 85-86-9 | C22H16N4O | 352.397 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA18318.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS19524-06-2
IUPAC Name4-bromopyridine hydrochloridyl
Molecular FormulaC5H4BrClN
InChI KeyPSPIMDWZDKGZBO-UHFFFAOYSA-N
SMILES[Cl].BrC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow-pink
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Appearance (Form)Powder