2-Fluorophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
2-Fluorophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
2-Fluorophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
2-Fluorophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Fluorophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
CAS: 38985-64-7 | C7H4FNS | 153.17 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18314.065 g
Catalog number ALFA18314.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Fluorophenyl isothiocyanate
CAS38985-64-7
Health Hazard 1H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c