2-Methyl-2-butanol, 98%, Thermo Scientific Chemicals
2-Methyl-2-butanol, 98%, Thermo Scientific Chemicals
2-Methyl-2-butanol, 98%, Thermo Scientific Chemicals
2-Methyl-2-butanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Methyl-2-butanol, 98%, Thermo Scientific Chemicals
CAS: 75-85-4 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
Catalog number ALFA18304.AP
Price (MYR)
528.00
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
528.00
Add to cart
Specifications
Chemical Name or Material2-Methyl-2-butanol
CAS75-85-4
Health Hazard 1H225-H312+H332-H315-H318-H335-H336
Health Hazard 2GHS H Statement
H225-H332-H315-H335
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
View more
2-Methyl-2-butanol is used as a solvent in flavors, pharmaceuticals, corrosion inhibitors, resins, gums, coating materials and in organic synthesis. It acts as a frothing agent (ore flotation), surfactant (petroleum recovery) and stabilizing agent. Further, it is used in processing aids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methyl-2-butanol is used as a solvent in flavors, pharmaceuticals, corrosion inhibitors, resins, gums, coating materials and in organic synthesis. It acts as a frothing agent (ore flotation), surfactant (petroleum recovery) and stabilizing agent. Further, it is used in processing aids.
Solubility
Miscible with alcohol, ether, benzene, chloroform, glycerol, oils and acetone.
Notes
Light sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Nikoshvili, L.; Shimanskaya, E.; Bykov, A.; Yuranov, I.; Kiwi-Minsker, L.; Sulman, E. Selective hydrogenation of 2-methyl-3-butyn-2-ol over Pd-nanoparticles stabilized in hypercrosslinked polystyrene: Solvent effect. Catal. Today 2015, 241, 179-188.
  2. Vernuccio, S.; Von Rohr, P. R.; Medlock, J. General Kinetic Modeling of the Selective Hydrogenation of 2-Methyl-3-butyn-2-ol over a Commercial Palladium-Based Catalyst. Ind. Eng. Chem. Res. 2015, 54 (46), 11543-11551.