Indium(III) nitrate hydrate, In 29% min., Thermo Scientific Chemicals
Indium(III) nitrate hydrate, In 29% min., Thermo Scientific Chemicals
Indium(III) nitrate hydrate, In 29% min., Thermo Scientific Chemicals
Indium(III) nitrate hydrate, In 29% min., Thermo Scientific Chemicals
Indium(III) nitrate hydrate, In 29% min., Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number ALFA18252.22
Price (MYR)
4,345.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
4,345.00
Add to cart
Specifications
Chemical Name or MaterialIndium(III) nitrate hydrate
Melting Point100°C (in 2H2O)
CAS207398-97-8
Health Hazard 1H272-H315-H319
Health Hazard 2GHS H Statement
H272-H315-H319
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
View more
Indium(III) nitrate hydrate acts as an oxidizing agent. It is used in the manufacture of coordinated polymer films which are used in the preparation of indium(III) oxide nanoparticles. It is used in the synthesis of other indium complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Indium(III) nitrate hydrate acts as an oxidizing agent. It is used in the manufacture of coordinated polymer films which are used in the preparation of indium(III) oxide nanoparticles. It is used in the synthesis of other indium complexes.
Solubility
Soluble in water, methanol and ethanol.
Notes
It is stable at room temperature. Hygroscopic. Incompatible with strong reducing agents and strong acids.
RUO – Research Use Only
General References:
  1. Selvakumar, D.; Dharmaraj, N.; Kumar, N. S.; Padaki, V. C. Preparation, Surface Morphology, and Optical Properties Of Indium(III) Oxide Nanoparticles by Thermolysis Of Indium-Poly(Vinyl Alcohol) Coordination Polymer. J. Coord. Chem. 2014, 67 (11), 1938-1947.
  2. Samanamú, C. R.; Richards, A. F. Indium(III) one-dimensional polymers with pyrazine and pyrazine, 2-carboxylic acid as the spacer molecules. Polyhedron 2007, 26 (4), 923-928.
  3. Wang, H.; Xu, X.; Li, X.; Zhang, J.; Li, C. Synthesis and sintering of indium tin oxide nanoparticles by citrate-nitrate combustion method. Rare Met. 2010, 29 (4), 355-360.