Butyraldehyde, 98+%, Thermo Scientific Chemicals
Butyraldehyde, 98+%, Thermo Scientific Chemicals
Butyraldehyde, 98+%, Thermo Scientific Chemicals
Butyraldehyde, 98+%, Thermo Scientific Chemicals
Fisherbrand
Butyraldehyde, 98+%, Thermo Scientific Chemicals
CAS: 123-72-8 | C4H8O | 72.11 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18243.AP500 mL
Catalog number ALFA18243.AP
Price (MYR)
372.00
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
372.00
Add to cart
Specifications
Chemical Name or MaterialButyraldehyde
CAS123-72-8
Health Hazard 1H225-H319
Health Hazard 2GHS H Statement
H225
Highly flammable liquid and vapour.
Health Hazard 3P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
View more
Butyraldehyde is used as an intermediate in the production of synthetic resins, rubber accelerators, tanning auxiliaries, perfumes, flavors, plasticizers and high molecular weight polymers. It acts as a chemical intermediate to prepare other chemicals such as 2-ethylhexanol, n-butanol, polyvinyl butyral (PVB), n-butyric acid and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Butyraldehyde is used as an intermediate in the production of synthetic resins, rubber accelerators, tanning auxiliaries, perfumes, flavors, plasticizers and high molecular weight polymers. It acts as a chemical intermediate to prepare other chemicals such as 2-ethylhexanol, n-butanol, polyvinyl butyral (PVB), n-butyric acid and pharmaceuticals.
Solubility
Miscible with water, ethanol, ether, ethyl acetate, acetone and toluene.
Notes
Air sensitive. Store in cool place. Incompatible with oxidizing agents, reducing agents, strong bases and acids.
RUO – Research Use Only
General References:
  1. Lee, S.; Varma, A. Aldol condensation of n-butyraldehyde in a biphasic stirred tank reactor: Experiments and models. AIChE Journal 2015, 61 (7), 2228-2239.
  2. Schlager, S.; Neugebauer, H.; Haberbauer, M.; Hinterberger, G.; Sariciftci, N. S. Direct Electrochemical Addressing of Immobilized Alcohol Dehydrogenase for the Heterogeneous Bioelectrocatalytic Reduction of Butyraldehyde to Butanol. ChemCatChem 2015, 7 (6), 967 - 971.