1-Hexanol, 99%, Thermo Scientific Chemicals
1-Hexanol, 99%, Thermo Scientific Chemicals
1-Hexanol, 99%, Thermo Scientific Chemicals
1-Hexanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Hexanol, 99%, Thermo Scientific Chemicals
CAS: 111-27-3 | C6H14O | 102.177 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18232.AP500 mL
Catalog number ALFA18232.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS86-96-4
IUPAC Name1,2,3,4-tetrahydroquinazoline-2,4-dione
Molecular FormulaC8H6N2O2
InChI KeySDQJTWBNWQABLE-UHFFFAOYSA-N
SMILESO=C1NC(=O)C2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Appearance (Color)White to cream or pink to gray
1-Hexanol is used as a precursor to plasticizers, chemical intermediate for pharmaceuticals, perfume esters and antiseptics. Further, it serves as a perturbing agent on actomyosin adenosine triphosphatease. In addition to this, it is used to modulate the function of actomyosin motor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Hexanol is used as a precursor to plasticizers, chemical intermediate for pharmaceuticals, perfume esters and antiseptics. Further, it serves as a perturbing agent on actomyosin adenosine triphosphatease. In addition to this, it is used to modulate the function of actomyosin motor.
Solubility
Miscible with ethanol, acetone, chloroform, ether, benzene. Slightly miscible with carbon tetrachloride and water.
Notes
Incompatible with strong acids, acid chlorides, acid anhydrides and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Semenov, K. N.; Regueira, T.; Fernández, J.; Charykov, N. A.; Murin, I. V. Pressure dependence of the solubility of light fullerenes in 1-hexanol from 298.15 K to 363.15 K. J. Mol. Liq. 2015, 209, 71-76.
  2. Patel, V.; Ray, D.; Singh, K.; Abezgauz, L.; Marangoni, G.; Aswal, V. K.; Bahadur, P. 1-Hexanol triggered structural characterization of the worm-like micelle to vesicle transitions in cetyltrimethylammonium tosylate solutions. RSC Adv. 2015, 5 (107), 87758-87768.