n-Tridecane, 98+%, Thermo Scientific Chemicals
n-Tridecane, 98+%, Thermo Scientific Chemicals
n-Tridecane, 98+%, Thermo Scientific Chemicals
n-Tridecane, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Tridecane, 98+%, Thermo Scientific Chemicals
CAS: 629-50-5 | C13H28 | 184.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA18220.36
Price (MYR)
2,149.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
2,149.00
Add to cart
Specifications
Chemical Name or Materialn-Tridecane
CAS629-50-5
Health Hazard 1H227-H304
Health Hazard 2GHS H Statement
H304
May be fatal if swallowed and enters airways.
Health Hazard 3P210-P280-P301+P310-P331-P370+P378q-P501c
View more
n-Tridecane is used as a distillation chaser. It is a component of various fuels and solvents used in industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
n-Tridecane is used as a distillation chaser. It is a component of various fuels and solvents used in industry.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Lukešová, M.; Zgardzinska, B.; Švajdlenková, H.; Zaleski, R.; Charmas, B.; Bartoš, J. Spin probe dynamics in relation to free volume in crystalline organics from ESR and PALS: N-tridecane. Physica B 2015, 476, 100-108.
  2. Sun, M.; Feng, J.; Bu, Y.; Wang, X.; Duan, H.; Luo, C. Graphene coating bonded onto stainless steel wire as a solid-phase microextraction fiber. Talanta 2015, 134, 200-205.