n-Heneicosane, 99%, Thermo Scientific Chemicals
n-Heneicosane, 99%, Thermo Scientific Chemicals
n-Heneicosane, 99%, Thermo Scientific Chemicals
n-Heneicosane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Heneicosane, 99%, Thermo Scientific Chemicals
CAS: 629-94-7 | C21H44 | 296.583 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFA18198.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Materialn-Heneicosane
CAS629-94-7
Melting Point39°C to 43°C
Recommended StorageAmbient temperatures
Density0.792