o-Tolidine dihydrochloride, 97%, Thermo Scientific Chemicals
o-Tolidine dihydrochloride, 97%, Thermo Scientific Chemicals
o-Tolidine dihydrochloride, 97%, Thermo Scientific Chemicals
o-Tolidine dihydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
o-Tolidine dihydrochloride, 97%, Thermo Scientific Chemicals
CAS: 612-82-8 | C14H18Cl2N2 | 285.212 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA18196.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Materialo-Tolidine dihydrochloride
CAS612-82-8
Health Hazard 1H302-H350
Health Hazard 2GHS H Statement
H350-H302
May cause cancer.
Harmful if swallowed.
Health Hazard 3P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
View more
o-Tolidine dihydrochloride is used as a sensitive colorimetric reagent for gold and for free chlorine in water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
o-Tolidine dihydrochloride is used as a sensitive colorimetric reagent for gold and for free chlorine in water.
Solubility
Soluble in water.
Notes
Air and light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. L.L.Salomon; J.E.Johnson. Enzymatic microdetermination of glucose in blood and urine. Anal. Chem. 1959, 31,(3), 453-456.
  2. A.N.Whitaker; M.J.Elms; P.P.Masci; P.G.Bundesen; D.B.Rylatt; A.J.Webber; I.H.Bunce. Measurement of cross linked fibrin derivatives in plasma: an immunoassay using monoclonal antibodies. J Clin Pathol. 1984, 37,(8), 882-887.