o-Tolidine dihydrochloride, 97%, Thermo Scientific Chemicals
o-Tolidine dihydrochloride, 97%, Thermo Scientific Chemicals
o-Tolidine dihydrochloride, 97%, Thermo Scientific Chemicals
o-Tolidine dihydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
o-Tolidine dihydrochloride, 97%, Thermo Scientific Chemicals
CAS: 612-82-8 | C14H18Cl2N2 | 285.212 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALFA18196.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS96-97-9
IUPAC Name2-hydroxy-5-nitrobenzoic acid
Molecular FormulaC7H5NO5
InChI KeyPPDRLQLKHRZIJC-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(=CC=C1O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Melting point228°C to 233°C
Appearance (Color)Off-white to yellow to beige to orange to tan
HPLC>=98.5 %
o-Tolidine dihydrochloride is used as a sensitive colorimetric reagent for gold and for free chlorine in water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
o-Tolidine dihydrochloride is used as a sensitive colorimetric reagent for gold and for free chlorine in water.
Solubility
Soluble in water.
Notes
Air and light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. L.L.Salomon; J.E.Johnson. Enzymatic microdetermination of glucose in blood and urine. Anal. Chem. 1959, 31,(3), 453-456.
  2. A.N.Whitaker; M.J.Elms; P.P.Masci; P.G.Bundesen; D.B.Rylatt; A.J.Webber; I.H.Bunce. Measurement of cross linked fibrin derivatives in plasma: an immunoassay using monoclonal antibodies. J Clin Pathol. 1984, 37,(8), 882-887.