Isonicotinamide, 99%, Thermo Scientific Chemicals
Isonicotinamide, 99%, Thermo Scientific Chemicals
Isonicotinamide, 99%, Thermo Scientific Chemicals
Isonicotinamide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isonicotinamide, 99%, Thermo Scientific Chemicals
CAS: 1453-82-3 | C6H6N2O | 122.127 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
50 g
Catalog number ALFA18190.22
Price (MYR)
510.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
510.00
Add to cart
Specifications
Chemical Name or MaterialIsonicotinamide
CAS1453-82-3
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c