Methylene Blue, high purity, biological stain
Methylene Blue, high purity, biological stain
Methylene Blue, high purity, biological stain
Methylene Blue, high purity, biological stain
Thermo Scientific Chemicals
Methylene Blue, high purity, biological stain
CAS: 122965-43-9 | C16H18ClN3S | 319.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ALFA18174.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS612-24-8
IUPAC Name2-nitrobenzonitrile
Molecular FormulaC7H4N2O2
InChI KeySWBDKCMOLSUXRH-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to pale yellow
Melting Point (clear melt)106.0-112.0?C
FormPowder
Assay (GC)≥98.0%
Methylene blue is used as a dye in different staining procedures viz. Wright's stain and Jenner's stain. It also serves as an indicator and medicine. Further, it is used to examine RNA and DNA under the microscope or in a gel. It is widely used as a redox indicator in analytical chemistry. It acts as a photosensitizer utilized to generate singlet oxygen in order to make organic peroxides by a Diels-Alder reaction. It is also used in the sulfide analysis. It plays an important role in aquaculture to treat fungal infections.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methylene blue is used as a dye in different staining procedures viz. Wright′s stain and Jenner′s stain. It also serves as an indicator and medicine. Further, it is used to examine RNA and DNA under the microscope or in a gel. It is widely used as a redox indicator in analytical chemistry. It acts as a photosensitizer utilized to generate singlet oxygen in order to make organic peroxides by a Diels-Alder reaction. It is also used in the sulfide analysis. It plays an important role in aquaculture to treat fungal infections.
Solubility
Soluble in water, alcohol, acetic acid, glycerol and chloroform. Insoluble in ether.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Chowdhury, P. R.; Bhattacharyya, K. G. Ni/Ti layered double hydroxide: synthesis, characterization and application as a photocatalyst for visible light degradation of aqueous methylene blue. Dalton Trans. 2015, 44 (15), 6809-6824.
  2. Das, M.; Bhattacharyya, K. G. Use of Raw and Acid-Treated MnO2 as Catalysts for Oxidation of Dyes in Water: A Case Study with Aqueous Methylene Blue. Chem. Eng. Commun. 2014, 202 (12), 1657-1667.