1,4-Dibromotetrafluorobenzene, 99%, Thermo Scientific Chemicals
1,4-Dibromotetrafluorobenzene, 99%, Thermo Scientific Chemicals
1,4-Dibromotetrafluorobenzene, 99%, Thermo Scientific Chemicals
1,4-Dibromotetrafluorobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Dibromotetrafluorobenzene, 99%, Thermo Scientific Chemicals
CAS: 344-03-6 | C6Br2F4 | 307.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFA18135.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material1,4-Dibromotetrafluorobenzene
CAS344-03-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
It is used as a chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a chemical intermediate.
Solubility
Solubility in methanol is almost transparent.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. A. Keith. Jameson.; Cynthia J. Jameson.Absolute temperature dependence of chemical shifts of lock solvents. Tetramethylsilane, hexafluorobenzene, and 1,4-dibromotetrafluorobenzene. Am. Chem. Soc.1973,95(26), 8559-8561.
  2. Alessandra Forni. Experimental and Theoretical Study of the Br···N Halogen Bond in Complexes of 1,4-Dibromotetrafluorobenzene with Dipyridyl Derivatives.J. Phys. Chem. A,2009,113(14), 3403-3412.