Lithium aluminum hydride, powder, 97%, Thermo Scientific™
Lithium aluminum hydride, powder, 97%, Thermo Scientific™
Lithium aluminum hydride, powder, 97%, Thermo Scientific™
Lithium aluminum hydride, powder, 97%, Thermo Scientific™
Lithium aluminum hydride, powder, 97%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFA18116.22100 g
Catalog number ALFA18116.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLithium aluminum hydride
Melting Point>125°C (decomposition)
CAS16853-85-3
Health Hazard 1H260-H314-H500
Health Hazard 2GHS H Statement
H260-H301-H314-H318
In contact with water releases flammable gases which may ignite spontaneously.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Lithium aluminum hydride is used as a reducing agent in various synthetic organic chemistry reactions. For example, it is used in the conversion of esters, carboxylic acids, acyl chlorides, aldehydes and ketones into their corresponding alcohols. It also involved in the conversion of amide, nitro, nitrile, imine, oxime and azide compounds into their amines as well as reduces alkyl halides to alkanes. It reacts with sodium chloride to prepare sodium hydride. It acts as a potential hydrogen storage medium, which could be used for future fuel cell-powered vehicles. It is mainly used in the preparation of other tetrahydridoaluminumates like sodium aluminum hydride, potassium aluminum hydride and magnesium alanate. Furthermore, it has been used to prepare thermoplastic polyester polyamides from oleic acid and lithium-polymer batteries.
Solubility
Soluble in water, diethyl ether and terahydrofuran.
Notes
Air and moisture sensitive. Incompatible with water, strong oxidizing agents, alcohols, carboxylic acid, peroxides, chlorinated solvents, halogens and oxygen. It reacts violently with water and release hydrogen gas.
General References:
  1. Many procedures for reactions with this reagent which have been described in the literature are potentially unsafe on anything other than a very small scale, since often no precautions are given for mixing the reagent with flammable solvents. The following suggestions are offered for laboratory scale reductions:
  2. Mixing with a flammable solvent must always be carried out under an inert atmosphere.
  3. Reverse addition (solvent to reagent), if done rapidly, may be safer.
  4. For THF, at least 10ml solvent per gram of reagent is recommended to absorb heat of mixing.
  5. Efficient stirring is essential during mixing, reaction and workup.
  6. The use of water or steam baths, or of electric heating mantles is to be strongly discouraged.
  7. The reduction reaction should preferably be controlled by adjusting the rate of addition of the substrate rather than relying on external heating or cooling.
  8. External heating or cooling, if required, should be by means of a suitable inert oil.
  9. The use of Diethoxymethane, B23134 as a reaction solvent has also been recommended.
  10. For phase-transfer reductions in hydrocarbon solvents, see 15-Crown-5, A12265. For formation of lithium tris(diethylamino)aluminum hydride and use in the reduction of esters or amides to aldehydes, see Diethyl amine, A11716. For formation of a reagent for reduction of carboxylic acids to aldehydes, see 1-Methyl piperazine, A10837.
  11. For reduction of sulfones, see Titanium(IV) chloride, 14713.
  12. Monograph: J. Seyden-Penne, Reductions by the Alumino- and Borohydrides In Organic Synthesis, 2nd ed., Wiley, N.Y. (1997).