Cadmium iodide, 98%, Thermo Scientific Chemicals
Cadmium iodide, 98%, Thermo Scientific Chemicals
Cadmium iodide, 98%, Thermo Scientific Chemicals
Cadmium iodide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cadmium iodide, 98%, Thermo Scientific Chemicals
CAS: 7790-80-9 | CdI2 | 366.223 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA18079.22
Price (MYR)
1,381.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,381.00
Add to cart
Specifications
Boiling Point787°C
Chemical Name or MaterialCadmium iodide
Melting Point387°C to 389°C
CAS7790-80-9
Health Hazard 1H301+H331-H351-H373
View more
It finds it use in photography, process engraving, lithography, analytical chemistry, and electroplating; Also used as in lubricants, phosphors. Used for cadmium electrode position and in photoconductors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds it use in photography, process engraving, lithography, analytical chemistry, and electroplating; Also used as in lubricants, phosphors. Used for cadmium electrode position and in photoconductors.
Solubility
Very soluble in water, slightly soluble in acetone, ethanol and diethyl ether.
Notes
Light Sensitive & Hygroscopic. Store away from oxidizing agents, light and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only
General References:
  1. Dipak Prajapati and Jagir S. Sandhu. Cadmium iodide as a new catalyst for knoevenagel condensations.J. Chem. Soc., Perkin Trans. 1.;1993,6739-740.
  2. J. L. Lambert. Linear Starch Reagents. Cadmium Iodide-Linear Starch Reagent.Anal. Chem.,1951,23(9), 1247-1251.