Magnesium carbonate hydroxide pentahydrate, light, 98%, Thermo Scientific Chemicals
Magnesium carbonate hydroxide pentahydrate, light, 98%, Thermo Scientific Chemicals
Magnesium carbonate hydroxide pentahydrate, light, 98%, Thermo Scientific Chemicals
Magnesium carbonate hydroxide pentahydrate, light, 98%, Thermo Scientific Chemicals
Magnesium carbonate hydroxide pentahydrate, light, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Magnesium carbonate hydroxide pentahydrate, light, 98%, Thermo Scientific Chemicals
CAS: 56378-72-4 | C4H12Mg5O19 | 485.65 g/mol
Have Questions?
Catalog NumberdimQuantity
ALFA18070.30
Catalog number ALFA18070.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight485.65 (395.58 Anhydrous)
Unit Size
Magnesium carbonate hydroxide pentahydrate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Magnesium carbonate hydroxide pentahydrate is used as pharmaceutical intermediate.
Solubility
Soluble in water and acids. Insoluble in alcohol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. M.Rigolo; R.T.Woodhams. Basic magnesium carbonate flame retardants for polypropylene. Polymer Engineering & Science. 1992, 32,(5), 327-334.
  2. Supon Ananta; Rik Brydson; Noel W.Thomas. Synthesis, formation and characterisation of MgNb2O6 powder in a columbite-like phase. Journal of the European Ceramic Society. 1999, 19,(3), 355-362 .