Dithizone, 98%, Thermo Scientific Chemicals
Dithizone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dithizone, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
50 g
250 g
Catalog number ALFA17947.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDithizone
CAS60-10-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Dithizone acts as an indicator for metals. It is also used as a chelating agent for heavy metals like mercury and lead. It is also used to evaluate the purification of human pancreatic islet preparations, which is used for transplantation in patients with type 1 diabetes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dithizone acts as an indicator for metals. It is also used as a chelating agent for heavy metals like mercury and lead. It is also used to evaluate the purification of human pancreatic islet preparations, which is used for transplantation in patients with type 1 diabetes.
Solubility
Soluble in ethanol, chloroform, dimethylsulfoxide, and pyridine. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Reagent for extraction of metals. Indicator for metals: Anal. Chim. Acta, 57, 393 (1971).
  2. Saxena, R.; Tiwari, S.; Sharma, N. Flow-injection solid phase extraction using Dowex Optipore L493 loaded with dithizone for preconcentration of chromium species from industrial waters and determination by FAAS. RSC Adv. 2015, 5 (85), 69196-69204.
  3. Zhang, L.; Xiong, Z.; Zhang, L.; Yu, B.; Zhang, W. Magnetic nanoparticles coated with dithizone-modified chitosan for use in solid-phase extraction of copper(II). Anal. Methods 2015, 7 (5), 2050-2054.