Malonamide, 98%, Thermo Scientific Chemicals
Malonamide, 98%, Thermo Scientific Chemicals
Malonamide, 98%, Thermo Scientific Chemicals
Malonamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Malonamide, 98%, Thermo Scientific Chemicals
CAS: 108-13-4 | C3H6N2O2 | 102.093 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17939.22100 g
Catalog number ALFA17939.22
Price (MYR)
360.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
360.00
Add to cart
Specifications
Chemical Name or MaterialMalonamide
CAS108-13-4
Melting Point167°C to 171°C
Recommended StorageAmbient temperatures
RTECS NumberON9125000
View more
Malonamide is a reagent for fluorimetric determination of reducing carbohydrates. The malonamide-based ionic liquid extractant was used in the extraction of europium(iii) and other trivalent rare earth elements from nitric acid medium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Malonamide is a reagent for fluorimetric determination of reducing carbohydrates. The malonamide-based ionic liquid extractant was used in the extraction of europium(iii) and other trivalent rare earth elements from nitric acid medium.
Solubility
Soluble in HCl.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Carmen E. Lisowski and James E. Hutchison. Malonamide-Functionalized Gold Nanoparticles for Selective, Colorimetric Sensing of Trivalent Lanthanide Ions. Anal. Chem. 2009, 81, (24), 10246-10253.
  2. C. Erlinger; L. Belloni; Th. Zemb; and C. Madic. Attractive Interactions between Reverse Aggregates and Phase Separation in Concentrated Malonamide Extractant Solutions. Langmuir. 1999, 15, (7), 2290-2300.
  3. Reagent for fluorimetric determination of reducing carbohydrates: Anal. Chim. Acta, 108, 421 (1979).