o-Tolidine, 98%, Thermo Scientific Chemicals
o-Tolidine, 98%, Thermo Scientific Chemicals
o-Tolidine, 98%, Thermo Scientific Chemicals
o-Tolidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
o-Tolidine, 98%, Thermo Scientific Chemicals
CAS: 119-93-7 | C14H16N2 | 212.296 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALFA17837.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS142-82-5
IUPAC Nameheptane
Molecular FormulaC7H16
InChI KeyIMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILESCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.03 %
Color scale=<10 APHA
UVat 220 nm A: =<0.15
UVat 240 nm A: =<0.01
UVat 210 nm A: =<0.4
View more
O-Tolidine is widely used in dye production. It is particularly used in textile, leather and paper industries. It can be used as an indicator in analytical, clinical and forensic chemistry. It is utilized for the determination of gold and chlorine level in swimming pool water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
O-Tolidine is widely used in dye production. It is particularly used in textile, leather and paper industries. It can be used as an indicator in analytical, clinical and forensic chemistry. It is utilized for the determination of gold and chlorine level in swimming pool water.
Solubility
Slightly soluble in water.
Notes
Light and air sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Kumar, A.; Sharma, K. K.; Kumar, P.; Ramchiary, N. Laccase isozymes from Ganoderma lucidum MDU-7: Isolation, characterization, catalytic properties and differential role during oxidative stress. J. Mol. Catal. B: Enzym. 2015, 113, 68-75.
  2. Kagalkar, A. N.; Khandare, R. V.; Govindwar, S. P. Textile dye degradation potential of plant laccase significantly enhances upon augmentation with redox mediators. RSC Adv. 2015, 5 (98), 80505-80517.