L-Ascorbic acid sodium salt, 99%, Thermo Scientific Chemicals
L-Ascorbic acid sodium salt, 99%, Thermo Scientific Chemicals
L-Ascorbic acid sodium salt, 99%, Thermo Scientific Chemicals
L-Ascorbic acid sodium salt, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L-Ascorbic acid sodium salt, 99%, Thermo Scientific Chemicals
CAS: 134-03-2 | C6H10NaO6 | 201.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA17759.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS1840-19-3
IUPAC Name1-isothiocyanato-3-(trifluoromethyl)benzene
Molecular FormulaC8H4F3NS
InChI KeyGFEPANUKFYVALF-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC(=C1)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to white to yellow
Assay (GC)≥97.5%
Refractive Index1.5530-1.5570 @ 20?C
FormLiquid
L-Ascorbic acid sodium salt is used as an antioxidant, an acidity regulator and an antimicrobial in food stuffs. As a reductant, it is involved in the isolation of chloroplasts. It acts as a coenzyme for many hydroxylation reactions. It plays a vital role to eliminate chronic and acute infections. It finds applications as an additive in curing meat products such as ham and sausage meat. It is widely used as a nutritional supplement in agriculture and animal feed to promote the growth of young animals and prevent scurvy. It is also used as an intermediate in the manufacture of various organic chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Ascorbic acid sodium salt is used as an antioxidant, an acidity regulator and an antimicrobial in food stuffs. As a reductant, it is involved in the isolation of chloroplasts. It acts as a coenzyme for many hydroxylation reactions. It plays a vital role to eliminate chronic and acute infections. It finds applications as an additive in curing meat products such as ham and sausage meat. It is widely used as a nutritional supplement in agriculture and animal feed to promote the growth of young animals and prevent scurvy. It is also used as an intermediate in the manufacture of various organic chemicals.
Solubility
Soluble in water. Slightly soluble in alcohol. Insoluble in chloroform and ether.
Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only
General References:
  1. Li, Q.; Wang, Z.; Zhang, S.; Zheng, W.; Zhao, Q.; Zhang, J.; Wang, L.; Wang, S.; Kong, D. Functionalization of the surface of electrospun poly (epsilon-caprolactone) mats using zwitterionic poly (carboxybetaine methacrylate) and cell-specific peptide for endothelial progenitor cells capture. Mater. Sci. Eng., C 2013, 33 (3), 1646-1653.
  2. de Tercero, M. D.; Martínez, I. G.; Herrmann, M.; Bruns, M.; Kübel, C.; Jennewein, S.; Fehrenbacher, U.; Barner, L.; Turk, M. Synthesis of in situ functionalized iron oxide nanoparticles presenting alkyne groups via a continuous process using near-critical and supercritical water. J. Supercrit. Fluids 2013, 82, 83-95.