1-Bromo-4-nitrobenzene, 98%, Thermo Scientific Chemicals
1-Bromo-4-nitrobenzene, 98%, Thermo Scientific Chemicals
1-Bromo-4-nitrobenzene, 98%, Thermo Scientific Chemicals
1-Bromo-4-nitrobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromo-4-nitrobenzene, 98%, Thermo Scientific Chemicals
CAS: 586-78-7 | C6H4BrNO2 | 202.01 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17705.065 g
Catalog number ALFA17705.06
Price (MYR)
259.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
259.00
Add to cart
Specifications
Chemical Name or Material1-Bromo-4-nitrobenzene
CAS586-78-7
Health Hazard 1H302+H312+H332-H315-H319-H335-H373
Health Hazard 2GHS H Statement
H331-H373-H302-H312-H315-H319-H335
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
View more
It can be employed as a syntheses material intermediate. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can be employed as a syntheses material intermediate. It is also used as a pharmaceutical intermediate.
Solubility
Insoluble in water. Almost transparent in hot ethyl alcohol. Soluble in ether
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry place.
RUO – Research Use Only
General References:
  1. Sven Ernst.; Sarah E Norman.; Christopher Hardacre.; Richard G Compton. The electrochemical reduction of 1-bromo-4-nitrobenzene at zinc electrodes in a room-temperature ionic liquid: a facile route for the formation of arylzinc compounds.Phys Chem Chem Phys.2014,16(10), 4478-4482.
  2. Jin-Heng Li.; Yun Liang.; De-Ping Wang.; Wen-Jie Liu.; Ye-Xiang Xie.; Du-Lin Yin. Efficient Stille cross-coupling reaction catalyzed by the Pd(OAc)2/Dabco catalytic system.J Org Chem.2005,70(7), 2832-2834.