2-Bromoethylamine hydrobromide, 98+%, Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 98+%, Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 98+%, Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 98+%, Thermo Scientific Chemicals
CAS: 2576-47-8 | C2H7Br2N | 204.893 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number ALFA17625.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Bromoethylamine hydrobromide
CAS2576-47-8
Health Hazard 1H302-H315-H317-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H317-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
View more
2-Bromoethylamine hydrobromide is used to construct C2-symmetric imidazolidinylidene ligands with a dioxolane backbone. It acts as a building block which is used for proteomics research. It is also used as organic and pharmaceutical intermediates for the preparation of first-aid medicines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromoethylamine hydrobromide is used to construct C2-symmetric imidazolidinylidene ligands with a dioxolane backbone. It acts as a building block which is used for proteomics research. It is also used as organic and pharmaceutical intermediates for the preparation of first-aid medicines.
Solubility
Soluble in water.
Notes
Hygroscopic and moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. O'Connor, L. J.; Cazares-Körner, C.; Saha, J.; Evans, C. N.; Stratford, M. R.; Hammond, E. M.; Conway, S. J. Efficient synthesis of 2-nitroimidazole derivatives and the bioreductive clinical candidate Evofosfamide (TH-302). Org. Chem. Front. 2015, 2 (9), 1026-1029.
  2. Maity, N.; Mandal, A.; Nandi, A. K. Interface engineering of ionic liquid integrated graphene in poly(vinylidene fluoride) matrix yielding magnificent improvement in mechanical, electrical and dielectric properties. Polymer 2015, 65, 154-167.