Barium perchlorate trihydrate, 99+%, Thermo Scientific™
Barium perchlorate trihydrate, 99+%, Thermo Scientific™
Barium perchlorate trihydrate, 99+%, Thermo Scientific™
Barium perchlorate trihydrate, 99+%, Thermo Scientific™
Barium perchlorate trihydrate, 99+%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFA17598.22100 g
Catalog number ALFA17598.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBarium perchlorate trihydrate
CAS10294-39-0
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H271-H302-H332
May cause fire or explosion; strong oxidizer.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3GHS P Statement
P221-P283-P210-P306+P360-P371+P380+P375-P501a
Take any precaution to avoid mixing with combustibles.
Wear fire/flame resistant/retardant clothing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON CLOTHING: rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
In case of major fire and large quantities: Evacuate area. Fight fire remotely due to the risk of explosion.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.