1-Tetradecylphosphonic acid, 98%, Thermo Scientific Chemicals
1-Tetradecylphosphonic acid, 98%, Thermo Scientific Chemicals
1-Tetradecylphosphonic acid, 98%, Thermo Scientific Chemicals
1-Tetradecylphosphonic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Tetradecylphosphonic acid, 98%, Thermo Scientific Chemicals
CAS: 4671-75-4 | C14H31O3P | 278.373 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17511.031 g
Catalog number ALFA17511.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Tetradecylphosphonic acid
CAS4671-75-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
1-Tetradecylphosphonic acid is used to functionalize the surface of BaTiO3 nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Tetradecylphosphonic acid is used to functionalize the surface of BaTiO3 nanoparticles.
Solubility
Soluble in ethanol, dimethyl sulfoxide and dimethyl formamide. Partially soluble in water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible withstrong oxidizing agents.
RUO – Research Use Only
General References:
  1. Ye, H. J.; Shao, W. Z.; Zhen, L. Tetradecylphosphonic acid modified BaTiO3 nanoparticles and its nanocomposite. Colloids Surf., A 2013, 427, 19-25.
  2. Wang, H.-r.; Jiang, Y.; Qu, J.-e.; Cao, Z.-y.; Liu, S.-b.; Guo, X.-p. Adsorption and Corrosion Inhibition Performances of 1-Tetradecylphosphonic Acid Self-Assembled Monolayers on 430 Stainless Steel Surface. J. Iron. Steel Res. Int. 2013, 20 (9), 93-98.