Creatine, anhydrous, 98%, Thermo Scientific Chemicals
Creatine, anhydrous, 98%, Thermo Scientific Chemicals
Creatine, anhydrous, 98%, Thermo Scientific Chemicals
Creatine, anhydrous, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Creatine, anhydrous, 98%, Thermo Scientific Chemicals
CAS: 57-00-1 | C4H9N3O2 | 131.135 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number ALFA17477.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCreatine
Name Noteanhydrous
CAS57-00-1
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Creatine is involved in the rapid production of ATP associated with creatine kinases in skeletal muscle tissue. Creatine may be used as a supplement to study its mechanism of action. It acts as a nutraceutical and a neuroprotective agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Creatine is involved in the rapid production of ATP associated with creatine kinases in skeletal muscle tissue. Creatine may be used as a supplement to study its mechanism of action. It acts as a nutraceutical and a neuroprotective agent.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Persky AM, et al. Clinical pharmacology of the dietary supplement creatine monohydrate.Pharmacological reviews.,2001,53(2), 161-176.
  2. Mendes RR, et al. [Creatine: the nutritional supplement for exercise - current concepts].Archivos latinoamericanos de nutricion.,2002,52(2), 117-127.