Amberlyst™ A-26(OH), ion exchange resin, Thermo Scientific Chemicals
Amberlyst™ A-26(OH), ion exchange resin, Thermo Scientific Chemicals
Amberlyst™ A-26(OH), ion exchange resin, Thermo Scientific Chemicals
Amberlyst™ A-26(OH), ion exchange resin, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Amberlyst™ A-26(OH), ion exchange resin, Thermo Scientific Chemicals
CAS: 39339-85-0
Have Questions?
Catalog NumberQuantity
ALFA17361.30250 g
Catalog number ALFA17361.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmberlyst™ A-26(OH)
Name Noteion exchange resin
CAS39339-85-0
Health Hazard 1H318
Health Hazard 3P280-P305+P351+P338-P310
View more
Amberlyst™ A-26(OH) is widely utilized to catalyze reactions such as aldol condensation and carbonylation. It is used to remove the anionic transition metal complexes from aqueous and non-aqueous mediums, acids and mercaptans from hydrocarbons and other non-polar solvents. It plays an essential role for the removal of oleic acid from chlorinated hydrocarbons and acids from phenol-acetone solutions. It is actively involved in the determination of cadmium by flow injection atomic absorption spectrometry and as packaging material for column chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Amberlyst|r A-26(OH) is widely utilized to catalyze reactions such as aldol condensation and carbonylation. It is used to remove the anionic transition metal complexes from aqueous and non-aqueous mediums, acids and mercaptans from hydrocarbons and other non-polar solvents. It plays an essential role for the removal of oleic acid from chlorinated hydrocarbons and acids from phenol-acetone solutions. It is actively involved in the determination of cadmium by flow injection atomic absorption spectrometry and as packaging material for column chromatography.
Solubility
Insoluble in water.
Notes
Exposure to air absorbs carbon dioxide. Incompatible with strong acids, bases and oxidants.
RUO – Research Use Only
General References:
  1. Macroreticular strongly basic anion excahnge resin, hydroxide form,suitable for aqueous and non-aqueous applications. Useful for removalof anionic metal complexes and mercaptans from non-polar solvents, andof acidic impurities from organic solutions.
  2. Haddad, R.; Thery, J.; Gauthier-Manuel, B.; Elouarzaki, K.; Holzinger, M.; Goff, A. L.; Gautier, G.; Mansouri, J. E.; Martinent, A.; Cosnier, S. High performance miniature glucose/O2 fuel cell based on porous silicon anion exchange membrane. Electrochem. Commun. 2015, 54, 10-13.
  3. Allen, C. E.; Curran, P. R.; Brearley, A. S.; Boissel, V.; Sviridenko, L.; Press, N. J.; Stonehouse, J. P.; Armstrong, A. Efficient and Facile Synthesis of Acrylamide Libraries for Protein-Guided Tethering. Org. Lett. 2015, 17 (3), 458-460.