1-Nitro-2-(n-octyloxy)benzene, 98%, Thermo Scientific Chemicals
1-Nitro-2-(n-octyloxy)benzene, 98%, Thermo Scientific Chemicals
1-Nitro-2-(n-octyloxy)benzene, 98%, Thermo Scientific Chemicals
1-Nitro-2-(n-octyloxy)benzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Nitro-2-(n-octyloxy)benzene, 98%, Thermo Scientific Chemicals
CAS: 37682-29-4 | C14H21NO3 | 251.326 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFA17320.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Nitro-2-(n-octyloxy)benzene
CAS37682-29-4
Recommended StorageAmbient temperatures
Density1.04
EINECS Number253-623-7